2-Bromo-3,4,5-trifluorobenzotrifluoride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-3,4,5-trifluoro-1-(trifluoromethyl)benzene, 1,2,3-Trifluoro-4-bromo-5-(trifluoromethyl)benzene

PC56735-1G PC56735-250MG
APOSPC56735-1GEA 835 EUR
APOSPC56735-1G APOSPC56735-250MG
2-Bromo-3,4,5-trifluorobenzotrifluoride 95%
2-Bromo-3,4,5-trifluorobenzotrifluoride
Formel: C₇HBrF₆
Molecular Weight: 278.977 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31009134
CAS-Nummer: 2088942-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR