3-Chloro-4,5-Difluorobenzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC56736-1G
PC56736-5G
APOSPC56736-1GEA
214
EUR
APOSPC56736-1G
APOSPC56736-5G
3-Chloro-4,5-Difluorobenzaldehyde 95%
3-Chloro-4,5-Difluorobenzaldehyde
Formel:
C₇H₃ClF₂O Molecular Weight: 176,548 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18415555 CAS-Nummer: 1261737-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...