Fluroxypyr-2-methoxy 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3,5-Dichloro-2-fluoro-6-methoxypyridin-4-amine
PC56755-1G
PC56755-250MG
APOSPC56755-1GEA
894
EUR
APOSPC56755-1G
APOSPC56755-250MG
Fluroxypyr-2-methoxy 95%
Fluroxypyr-2-methoxy
Formel:
C₆H₅Cl₂FN₂O Molecular Weight: 211,021 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19441410 CAS-Nummer: 35622-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...