4-(Fluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Gefahr

PC56766-1G PC56766-250MG
APOSPC56766-1GEA 493 EUR
APOSPC56766-1G APOSPC56766-250MG
4-(Fluoromethyl)benzoic acid 95%
4-(Fluoromethyl)benzoic acid
Formel: C₈H₇FO₂
Molecular Weight: 154.138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20482169
CAS-Nummer: 118507-45-2
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR