2-Chloro-4-(trifluoromethoxy)benzonitrile 95%

Lieferant: Apollo Scientific

PC56770-1G PC56770-250MG PC56770-5G
APOSPC56770-1GEA 870 EUR
APOSPC56770-1G APOSPC56770-250MG APOSPC56770-5G
2-Chloro-4-(trifluoromethoxy)benzonitrile 95%
2-Chloro-4-(trifluoromethoxy)benzonitrile
Formel: C₈H₃ClF₃NO
Molecular Weight: 221.564 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18393803
CAS-Nummer: 874482-94-7
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR