2-Chloro-4-(trifluoromethoxy)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC56771-1G PC56771-5G
APOSPC56771-1GEA 870 EUR
APOSPC56771-1G APOSPC56771-5G
2-Chloro-4-(trifluoromethoxy)benzoic acid 95%
2-Chloro-4-(trifluoromethoxy)benzoic acid
Formel: C₈H₄ClF₃O₃
Molecular Weight: 240.564 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18393797
CAS-Nummer: 1261731-19-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR