2-Nitrobenzyl fluoride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC56784-1G
PC56784-250MG
APOSPC56784-1GEA
609
EUR
APOSPC56784-1G
APOSPC56784-250MG
2-Nitrobenzyl fluoride 95%
2-Nitrobenzyl fluoride
Formel:
C₇H₆FNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22053300 CAS-Nummer: 447-58-5 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...