4-Nitrobenzyl fluoride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC56788-1G
PC56788-250MG
APOSPC56788-1GEA
69.6
EUR
APOSPC56788-1G
APOSPC56788-250MG
4-Nitrobenzyl fluoride 95%
4-Nitrobenzyl fluoride
Formel:
C₇H₆FNO₂ Molecular Weight: 155,126 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28990107 CAS-Nummer: 500-11-8 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...