6-Bromo-3-(difluoromethyl)-1H-indole, N-BOC protected

Lieferant: Apollo Scientific
Gefahr

Synonyme: tert-butyl 6-bromo-3-(difluoromethyl)indole-1-carboxylate

PC56795-100MG PC56795-1G PC56795-500MG
APOSPC56795-100MGEA 522 EUR
APOSPC56795-100MG APOSPC56795-1G APOSPC56795-500MG
6-Bromo-3-(difluoromethyl)-1H-indole, N-BOC protected
6-Bromo-3-(difluoromethyl)-1H-indole, N-BOC protected
Formel: C₁₄H₁₄BrF₂NO₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2237235-37-7
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR