3-Fluoro-1H-pyrazole 95%

Lieferant: Apollo Scientific
Achtung

PC56796-1G PC56796-250MG PC56796-100MG
APOSPC56796-1GEA 1390 EUR
APOSPC56796-1G APOSPC56796-250MG APOSPC56796-100MG
3-Fluoro-1H-pyrazole 95%
3-Fluoro-1H-pyrazole
Formel: C₃H₃FN₂
Molecular Weight: 86,068 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18205877
CAS-Nummer: 14521-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR