2,3,4-Trifluoro-6-(trifluoromethyl)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(Bromomethyl)-3,4,5-trifluoro-1-(trifluoromethyl)benzene

PC56797-1G PC56797-250MG
APOSPC56797-1GEA 654 EUR
APOSPC56797-1G APOSPC56797-250MG
2,3,4-Trifluoro-6-(trifluoromethyl)benzyl bromide 95%
2,3,4-Trifluoro-6-(trifluoromethyl)benzyl bromide
Formel: C₈H₃BrF₆
Molecular Weight: 293,004 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31634826
CAS-Nummer: 2244085-70-7
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR