2-Bromo-4'-(trifluoromethyl)-1,1'-biphenyl 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-[4-(trifluoromethyl)phenyl]benzene
PC56803-1G
PC56803-250MG
APOSPC56803-1GEA
1150
EUR
APOSPC56803-1G
APOSPC56803-250MG
2-Bromo-4'-(trifluoromethyl)-1,1'-biphenyl 95%
2-Bromo-4'-(trifluoromethyl)-1,1'-biphenyl
Formel:
C₁₃H₈BrF₃ Molecular Weight: 301,102 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03412188 CAS-Nummer: 255837-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...