2-(Difluoromethoxy)-5-nitropyridine 95%

Lieferant: Apollo Scientific
Achtung

PC56808-1G PC56808-5G PC56808-10G
APOSPC56808-1GEA 174 EUR
APOSPC56808-1G APOSPC56808-5G APOSPC56808-10G
2-(Difluoromethoxy)-5-nitropyridine 95%
2-(Difluoromethoxy)-5-nitropyridine
Formel: C₆H₄F₂N₂O₃
Molecular Weight: 190.104 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847317
CAS-Nummer: 1192813-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR