7-Fluorobenzo[b]thiophene-2-carbonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 7-Fluoro-1-benzothiophene-2-carbonyl chloride

PC56818-1G PC56818-250MG
APOSPC56818-1GEA 284 EUR
APOSPC56818-1G APOSPC56818-250MG
7-Fluorobenzo[b]thiophene-2-carbonyl chloride
7-Fluorobenzo[b]thiophene-2-carbonyl chloride
Formel: C₉H₄ClFOS
Molecular Weight: 214,644 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29477441
CAS-Nummer: 2035458-56-9
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR