3-Chloro-5-(trifluoromethoxy)-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC56830-1G PC56830-250MG
APOSPC56830-1GEA 522 EUR
APOSPC56830-1G APOSPC56830-250MG
3-Chloro-5-(trifluoromethoxy)-1H-indazole
3-Chloro-5-(trifluoromethoxy)-1H-indazole
Formel: C₈H₄ClF₃N₂O
Molecular Weight: 236,578 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29111399
CAS-Nummer: 1446406-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR