2-Bromo-5-fluorophenylacetylene 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-2-ethynyl-4-fluorobenzene

PC56836-1G PC56836-250MG PC56836-5G
APOSPC56836-1GEA 58 EUR
APOSPC56836-1G APOSPC56836-250MG APOSPC56836-5G
2-Bromo-5-fluorophenylacetylene 96%
2-Bromo-5-fluorophenylacetylene
Formel: C₈H₄BrF
Molecular Weight: 199.02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29046530
CAS-Nummer: 1693887-36-3
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR