2-Bromo-4-(trifluoromethyl)phenylacetylene 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-1-ethynyl-4-(trifluoromethyl)benzene
PC56840-1G
PC56840-250MG
APOSPC56840-1GEA
574
EUR
APOSPC56840-1G
APOSPC56840-250MG
2-Bromo-4-(trifluoromethyl)phenylacetylene 95%
2-Bromo-4-(trifluoromethyl)phenylacetylene
Formel:
C₉H₄BrF₃ Molecular Weight: 249,027 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD31689284 CAS-Nummer: 871126-88-4 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...