3-Bromo-2,4-difluorobenzoyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

PC56856-1G PC56856-250MG PC56856-5G
APOSPC56856-1GEA 493 EUR
APOSPC56856-1G APOSPC56856-250MG APOSPC56856-5G
3-Bromo-2,4-difluorobenzoyl chloride 95%
3-Bromo-2,4-difluorobenzoyl chloride
Formel: C₇H₂BrClF₂O
Molecular Weight: 255,444 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28737908
CAS-Nummer: 1507667-28-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR