4-[(4-Fluorophenyl)-hydroxymethyl]benzonitrile 98%

Lieferant: Apollo Scientific
Achtung

PC56868-1G PC56868-25G PC56868-5G
APOSPC56868-1GEA 92.8 EUR
APOSPC56868-1G APOSPC56868-25G APOSPC56868-5G
4-[(4-Fluorophenyl)-hydroxymethyl]benzonitrile 98%
4-[(4-Fluorophenyl)-hydroxymethyl]benzonitrile
Formel: C₁₄H₁₀FNO
Molecular Weight: 227,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11096849
CAS-Nummer: 220583-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR