6-(Trifluoromethyl)imidazo[1,5-a]pyrimidine-8-carbonitrile 95%

Lieferant: Apollo Scientific
Achtung

PC56880-1G PC56880-250MG
APOSPC56880-1GEA 609 EUR
APOSPC56880-1G APOSPC56880-250MG
6-(Trifluoromethyl)imidazo[1,5-a]pyrimidine-8-carbonitrile 95%
6-(Trifluoromethyl)imidazo[1,5-a]pyrimidine-8-carbonitrile
Formel: C₈H₃F₃N₄
Molecular Weight: 212,131 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31689288
CAS-Nummer: 2151834-44-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR