2-(Fluoromethyl)aniline, N-BOC protected 95%

Lieferant: Apollo Scientific
Gefahr

PC56900-1G PC56900-250MG
APOSPC56900-1GEA 567 EUR
APOSPC56900-1G APOSPC56900-250MG
2-(Fluoromethyl)aniline, N-BOC protected 95%
2-(Fluoromethyl)aniline, N-BOC protected
Formel: C₁₂H₁₆FNO₂
Molecular Weight: 225.259 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31664917
CAS-Nummer: 1374329-41-5
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR