7-(Trifluoromethyl)benzo[D]isoxazol-3-amine

Lieferant: Apollo Scientific
Gefahr

PC56915-1G PC56915-250MG
APOSPC56915-1GEA 306 EUR
APOSPC56915-1G APOSPC56915-250MG
7-(Trifluoromethyl)benzo[D]isoxazol-3-amine
7-(Trifluoromethyl)benzo[D]isoxazol-3-amine
Formel: C₈H₅F₃N₂O
Molecular Weight: 202,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19441875
CAS-Nummer: 229623-52-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR