3-Fluoro-4-chlorophenacyl bromide 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(4-chloro-3-fluorophenyl)ethanone

PC56972-1G PC56972-5G PC56972-25G
APOSPC56972-1GEA 159 EUR
APOSPC56972-1G APOSPC56972-5G APOSPC56972-25G
3-Fluoro-4-chlorophenacyl bromide 98%
3-Fluoro-4-chlorophenacyl bromide
Formel: C₈H₅BrClFO
Molecular Weight: 251.48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194237
CAS-Nummer: 231297-62-4
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR