[7-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol

Lieferant: Apollo Scientific
Achtung

PC56973-1G PC56973-250MG
APOSPC56973-1GEA 126 EUR
APOSPC56973-1G APOSPC56973-250MG
[7-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol
[7-(Trifluoromethyl)-1-benzothiophen-2-yl]methanol
Formel: C₁₀H₇F₃OS
Molecular Weight: 232.222 g/mol
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12032283
CAS-Nummer: 1171926-67-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR