(2S)-1-Fluoro-3-phenylpropan-2-amine, N-BOC protected 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl (S)-(1-fluoro-3-phenylpropan-2-yl)carbamate

PC56982-1G PC56982-250MG
APOSPC56982-1GEA 58 EUR
APOSPC56982-1G APOSPC56982-250MG
(2S)-1-Fluoro-3-phenylpropan-2-amine, N-BOC protected 95%
(2S)-1-Fluoro-3-phenylpropan-2-amine, N-BOC protected
Formel: C₁₄H₂₀FNO₂
Molecular Weight: 253,313 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30378947
CAS-Nummer: 2006287-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR