5-(Trifluoromethoxy)-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC57007-1G PC57007-5G PC57007-250MG
APOSPC57007-1GEA 168 EUR
APOSPC57007-1G APOSPC57007-5G APOSPC57007-250MG
5-(Trifluoromethoxy)-1H-indazole
5-(Trifluoromethoxy)-1H-indazole
Formel: C₈H₅F₃N₂O
Molecular Weight: 202,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16631268
CAS-Nummer: 105391-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR