3-Bromo-5-(trifluoromethoxy)-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC57009-1G PC57009-250MG
APOSPC57009-1GEA 836 EUR
APOSPC57009-1G APOSPC57009-250MG
3-Bromo-5-(trifluoromethoxy)-1H-indazole
3-Bromo-5-(trifluoromethoxy)-1H-indazole
Formel: C₈H₄BrF₃N₂O
Molecular Weight: 281,029 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18074536
CAS-Nummer: 1346521-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR