Methyl 2-bromo-5-fluoroisonicotinate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-5-fluoro-4-(methoxycarbonyl)pyridine, Methyl 2-bromo-5-fluoropyridine-4-carboxylate
PC57012-250MG
PC57012-1G
PC57012-5G
APOSPC57012-250MGEA
21.9
EUR
APOSPC57012-250MG
APOSPC57012-1G
APOSPC57012-5G
Methyl 2-bromo-5-fluoroisonicotinate 95%
Methyl 2-bromo-5-fluoroisonicotinate
Formel:
C₇H₅BrFNO₂ Molecular Weight: 234,023 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09951945 CAS-Nummer: 885588-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...