Methyl 6-chloro-5-fluoronicotinate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-3-fluoro-5-(methoxycarbonyl)pyridine, Methyl 6-chloro-5-fluoropyridine-3-carboxylate
PC57018-1G
APOSPC57018-1GEA
539
EUR
APOSPC57018-1G
Methyl 6-chloro-5-fluoronicotinate
Methyl 6-chloro-5-fluoronicotinate
Formel:
C₇H₅ClFNO₂ Molecular Weight: 189,572 g/mol Schmelzpunkt: 87…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD14698136 CAS-Nummer: 78686-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...