3-Fluoro-4-iodobenzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Fluoro-4-iodophenyl)methanol
PC57052-1G
PC57052-5G
PC57052-10G
APOSPC57052-1GEA
71.2
EUR
APOSPC57052-1G
APOSPC57052-5G
APOSPC57052-10G
3-Fluoro-4-iodobenzyl alcohol
3-Fluoro-4-iodobenzyl alcohol
Formel:
C₇H₆FIO Molecular Weight: 252,025 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18396947 CAS-Nummer: 1261615-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...