2-Nitrobenzal fluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Difluoromethyl)nitrobenzene, 1-(Difluoromethyl)-2-nitrobenzene, α,α-Difluoro-2-nitrotoluene

PC57055-1G PC57055-5G
APOSPC57055-1GEA 69.6 EUR
APOSPC57055-1G APOSPC57055-5G
2-Nitrobenzal fluoride
2-Nitrobenzal fluoride
Formel: C₇H₅F₂NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD23382510
CAS-Nummer: 64747-65-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR