4’-Bromo-2’-hydroxy-2,2,2-trifluoroacetophenone 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Bromo-2-hydroxyphenyl)-2,2,2-trifluoroethanone
PC57073-1G
PC57073-250MG
APOSPC57073-1GEA
140
EUR
APOSPC57073-1G
APOSPC57073-250MG
4’-Bromo-2’-hydroxy-2,2,2-trifluoroacetophenone 99%
4’-Bromo-2’-hydroxy-2,2,2-trifluoroacetophenone
Formel:
C₈H₄BrF₃O₂ Molecular Weight: 269.015 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31735860 CAS-Nummer: 1184850-45-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...