4-Bromo-3-chloro-2-fluorobenzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC57077-1G
PC57077-250MG
PC57077-5G
APOSPC57077-1GEA
290
EUR
APOSPC57077-1G
APOSPC57077-250MG
APOSPC57077-5G
4-Bromo-3-chloro-2-fluorobenzyl bromide 95%
4-Bromo-3-chloro-2-fluorobenzyl bromide
Formel:
C₇H₄Br₂ClF Molecular Weight: 302.366 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31922519 CAS-Nummer: 2091710-10-8 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...