3-Fluoro-4-iodo-2-methoxyaniline 95%

Lieferant: Apollo Scientific
Achtung

PC57083-1G PC57083-250MG
APOSPC57083-1GEA 719 EUR
APOSPC57083-1G APOSPC57083-250MG
3-Fluoro-4-iodo-2-methoxyaniline 95%
3-Fluoro-4-iodo-2-methoxyaniline
Formel: C₇H₇FINO
Molecular Weight: 267,039 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31654715
CAS-Nummer: 2092595-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR