4-Bromo-N-(2,2,2-trifluoroethyl)aniline
Lieferant: Apollo Scientific
|
Achtung
|
PC57088-1G
PC57088-250MG
APOSPC57088-1GEA
339
EUR
APOSPC57088-1G
APOSPC57088-250MG
4-Bromo-N-(2,2,2-trifluoroethyl)aniline
4-Bromo-N-(2,2,2-trifluoroethyl)aniline
Formel:
C₈H₇BrF₃N Molecular Weight: 254,047 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11153109 CAS-Nummer: 50885-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...