2-Fluoro-4-methyl-5-nitrobenzonitrile

Lieferant: Apollo Scientific
Gefahr

PC57102-1G PC57102-250MG PC57102-5G
APOSPC57102-1GEA 91.5 EUR
APOSPC57102-1G APOSPC57102-250MG APOSPC57102-5G
2-Fluoro-4-methyl-5-nitrobenzonitrile
2-Fluoro-4-methyl-5-nitrobenzonitrile
Formel: C₈H₅FN₂O₂
Molecular Weight: 180.136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22200940
CAS-Nummer: 1695920-54-7
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR