Methyl 2-bromo-6-fluoronicotinate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-bromo-6-fluoropyridine-3-carboxylate

PC57105-1G PC57105-250MG
APOSPC57105-1GEA 870 EUR
APOSPC57105-1G APOSPC57105-250MG
Methyl 2-bromo-6-fluoronicotinate 95%
Methyl 2-bromo-6-fluoronicotinate
Formel: C₇H₅BrFNO₂
Molecular Weight: 234,023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698113
CAS-Nummer: 1214385-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR