Diphenyl[4-(phenylthio)phenyl]sulphonium triflate

Lieferant: Apollo Scientific

PC5764-1G PC5764-5G
APOSPC5764-1GEA 82.8 EUR
APOSPC5764-1G APOSPC5764-5G
Diphenyl[4-(phenylthio)phenyl]sulphonium triflate
Diphenyl[4-(phenylthio)phenyl]sulphonium triflate

Photoinitiator/Photoacid Generator

Formel: C₂₄H₁₉S₂·CF₃O₃S
Molecular Weight: 520,607 g/mol
Schmelzpunkt: 81…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683569
CAS-Nummer: 111281-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR