6-(Trifluoromethyl)benzothiophene-2-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
PC57783-1G
PC57783-250MG
APOSPC57783-1GEA
157
EUR
APOSPC57783-1G
APOSPC57783-250MG
6-(Trifluoromethyl)benzothiophene-2-carbonitrile
6-(Trifluoromethyl)benzothiophene-2-carbonitrile
Formel:
C₁₀H₄F₃NS Molecular Weight: 227.206 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31567847 CAS-Nummer: 2121264-35-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...