2-(Bromomethyl)-6-(trifluoromethyl)benzothiophene

Lieferant: Apollo Scientific
Gefahr

PC57785-100MG PC57785-1G PC57785-250MG
APOSPC57785-100MGEA 33.5 EUR
APOSPC57785-100MG APOSPC57785-1G APOSPC57785-250MG
2-(Bromomethyl)-6-(trifluoromethyl)benzothiophene
2-(Bromomethyl)-6-(trifluoromethyl)benzothiophene
Formel: C₁₀H₆BrF₃S
Molecular Weight: 295.119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31567845
CAS-Nummer: 2377846-67-6
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR