5-(Trifluoromethyl)benzothiophene-2-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
PC57788-1G
PC57788-250MG
APOSPC57788-1GEA
136
EUR
APOSPC57788-1G
APOSPC57788-250MG
5-(Trifluoromethyl)benzothiophene-2-carbonitrile
5-(Trifluoromethyl)benzothiophene-2-carbonitrile
Formel:
C₁₀H₄F₃NS Molecular Weight: 227.206 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31567859 CAS-Nummer: 2121705-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...