[4-(Trifluoromethyl)benzothiophen-2-yl]methanol

Lieferant: Apollo Scientific
Achtung

PC57792-100MG PC57792-1G PC57792-250MG
APOSPC57792-100MGEA 21.9 EUR
APOSPC57792-100MG APOSPC57792-1G APOSPC57792-250MG
[4-(Trifluoromethyl)benzothiophen-2-yl]methanol
[4-(Trifluoromethyl)benzothiophen-2-yl]methanol
Formel: C₁₀H₇F₃OS
Molecular Weight: 232.222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12032277
CAS-Nummer: 1171926-60-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR