2,2,2-Trifluoro-N-{2-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenyl]ethyl}acetamide 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2,2-trifluoro-N-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenethyl)acetamide

PC57795-1G PC57795-250MG PC57795-5G
APOSPC57795-1GEA 37.7 EUR
APOSPC57795-1G APOSPC57795-250MG APOSPC57795-5G
2,2,2-Trifluoro-N-{2-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenyl]ethyl}acetamide 95%
2,2,2-Trifluoro-N-{2-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenyl]ethyl}acetamide
Formel: C₁₇H₂₀BF₆NO₃
Molecular Weight: 411,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32177068
CAS-Nummer: 2135636-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR