(1Z)-1-Bromo-2,4,4,4-tetrafluorobut-1-ene

Lieferant: Apollo Scientific
Achtung

PC5804-1G PC5804-5G
APOSPC5804-1GEA 129 EUR
APOSPC5804-1G APOSPC5804-5G
(1Z)-1-Bromo-2,4,4,4-tetrafluorobut-1-ene
(1Z)-1-Bromo-2,4,4,4-tetrafluorobut-1-ene
Formel: C₄H₃BrF₄
Molecular Weight: 206,964 g/mol
Dichte: >1,5 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD12026486
CAS-Nummer: 933600-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR