1-Iodo-2H-perfluoropropane 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2H-Perfluoropropyl iodide, 1-Iodo-1,1,2,3,3,3-hexafluoropropane

PC5821-1G
APOSPC5821-1GEA 306 EUR
APOSPC5821-1G
1-Iodo-2H-perfluoropropane 97%
1-Iodo-2H-perfluoropropane
Formel: C₃HF₆I
Molecular Weight: 277,935 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00069130
CAS-Nummer: 431-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR