2-(Iodomethyl)-1,1,1,3,3,3-hexafluoropropane
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,1,1,3,3,3-Hexafluoro-2-(iodomethyl)propane, 2H-Hexafluoro-2-(iodomethyl)propane
PC5826-1G
PC5826-25G
PC5826-5G
APOSPC5826-1GEA
94.5
EUR
APOSPC5826-1G
APOSPC5826-25G
APOSPC5826-5G
2-(Iodomethyl)-1,1,1,3,3,3-hexafluoropropane
2-(Iodomethyl)-1,1,1,3,3,3-hexafluoropropane
Formel:
C₄H₃F₆I Molecular Weight: 291,962 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038313 CAS-Nummer: 883449-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...