3-Chloro-4-iodo-2H,2H,4H,4H-perfluorobutane
Lieferant: Apollo Scientific
Synonyme:
3-Chloro-4-iodo-1,1,1,3-tetrafluorobutane
PC5828-1G
PC5828-5G
APOSPC5828-1GEA
157
EUR
APOSPC5828-1G
APOSPC5828-5G
3-Chloro-4-iodo-2H,2H,4H,4H-perfluorobutane
3-Chloro-4-iodo-2H,2H,4H,4H-perfluorobutane
Formel:
C₄H₄ClF₄I Molecular Weight: 290,426 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07784198 CAS-Nummer: 885275-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...