5-(Bromomethyl)-2-fluoropyridine

Lieferant: Apollo Scientific
Achtung

PC5845-250MG PC5845-100MG
APOSPC5845-250MGEA 0 EUR
APOSPC5845-250MG APOSPC5845-100MG
5-(Bromomethyl)-2-fluoropyridine
5-(Bromomethyl)-2-fluoropyridine
Formel: C₆H₅BrFN
Lagertemperatur: Kühlschrank
MDL: MFCD11520696
CAS-Nummer: 105827-74-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR