2-Bromo-6-(trifluoromethyl)phenylacetic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-2-(carboxymethyl)benzotrifluoride
PC5852-1G
PC5852-250MG
PC5852-100MG
APOSPC5852-1GEA
774
EUR
APOSPC5852-1G
APOSPC5852-250MG
APOSPC5852-100MG
2-Bromo-6-(trifluoromethyl)phenylacetic acid 98%
2-Bromo-6-(trifluoromethyl)phenylacetic acid
Formel:
C₉H₆BrF₃O₂ Molecular Weight: 283.042 g/mol Schmelzpunkt: 150…151 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026462 CAS-Nummer: 1159512-50-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...