1-(2-Fluorophenyl)propylamine 97%
Lieferant: Apollo Scientific
Synonyme:
1-(2-Fluorophenyl)propan-1-amine
PC5883-1G
APOSPC5883-1GEA
347
EUR
APOSPC5883-1G
1-(2-Fluorophenyl)propylamine 97%
1-(2-Fluorophenyl)propylamine
Formel:
C₉H₁₂FN Molecular Weight: 153,197 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07784283 CAS-Nummer: 473249-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...